Learn More
iQ Biosciences Purified Cynomolgus Monkey CD8 T Cells, 2M cells/vial
Supplier:  iQ Biosciences IQBMN1CD8T2
Encompass_Preferred
High-quality, high-viability (> 90%) CD8 T cells purified from cynomolgus (cyno) monkey (or macaque) PBMCs by magnetic selection. Highly enriched (typically > 90%) post-selection purity achieved via magnetic isolation. Used for a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.The following donor requests can be accommodated if inventory is available: - Single or multiple donors for a particular product (SKU). Indicate number of vials from each individual donor - Donor-matched PBMCs
Catalog No. NC2855567