Learn More
iQ Biosciences Purified Human CD8 T Cells, 10M cells/vial
Supplier:  iQ Biosciences IQBHU1CD8T10
Encompass_Preferred
High-quality, high-viability (> 90%) human CD8 T cells are purified from PBMCs of healthy blood donors sourced from a world-renowned blood center. Highly enriched (typically > 90%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.
Catalog No. NC1720477