Learn More
iQ Biosciences Purified Human Monocytes, 10M cells/vial
Supplier:  iQ Biosciences IQBHU1M10
Encompass
High-quality, high-viability (> 90%) human monocytes purified from PBMCs of healthy blood donors sourced from a world-renowned blood center. Highly enriched (> 90%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.
Catalog No. NC1911983