Learn More
iQ Biosciences Purified SD Rat CD3 Pan T Cells, 5M cells/vial
Supplier:  iQ Biosciences IQBR1T5
Encompass_Preferred
High-quality, high-viability (typically > 70%) CD3 Pan T cells purified from Sprague Dawley rat PBMCs. Highly enriched (> 95%) post-selection purity achieved via magnetic isolation. Used for a wide variety of safety assessment and functional assays for research and nonclinical drug development. All orders come with an iQ Certificate of Analysis.
Catalog No. NC2125133