Learn More
Medchemexpress LLC Purine (7H-Imidazo(4,5-d)pyrimidine) | 120-73-0 | 99.9% | 120.11 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY34431500MG
Encompass_Preferred
Purine is an endogenous metabolite and a fundamental building block of nucleic acids. It has demonstrated an ability to inhibit the activation of PARP, offering protection against oxidant-induced cell injury.
  • Inhibits PARP activation
  • Protects against oxidant-induced cell injury
  • Building block of nucleic acids
  • Suitable for research in cancer and nervous system diseases
Catalog No. 50-004-03655