Learn More
Medchemexpress LLC Purinostat mesylate | 2650188-32-0 | 99.45% | 586.62 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15010950MG
Encompass_Preferred
Purinostat mesylate is a selective inhibitor of HDACs, specifically inhibiting class I and class IIb HDACs. It is suitable for research related to lymphoblastic leukemia, demonstrating potent anti-leukemia effects in vivo by inducing apoptosis and affecting the cell cycle of LAMA84 and 188 BL-2 cells.
  • Selective inhibitor of class I and class IIb HDACs
  • Induces apoptosis in LAMA84 and 188 BL-2 cells
  • Affects cell cycle progression
  • Demonstrates potent anti-leukemia effects in vivo
  • Useful for research on lymphoblastic leukemia
Catalog No. 50-003-61323