Learn More
Medchemexpress LLC Purmorphamine | 483367-10-8 | C31H32N6O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15108100MG
Encompass_Preferred
Purmorphamine is a potent smoothened/Smo receptor agonist with an EC50 of 1 μM and an IC50 of 1.5 μM for Smoothened. It is intended for research use only and not for human or animal therapeutic use. This compound promotes osteoblast differentiation and up-regulates key markers of the osteoblast phenotype, demonstrating its utility in studying bone development and related cellular processes.
  • Acts as a smoothened/Smo receptor agonist
  • Induces osteoblast differentiation
  • Up-regulates alkaline phosphatase activity and RUNX-2 expression
  • Up-regulates osteocalcin at specific doses
  • Induces STAT3 phosphorylation
  • Promotes bone-like nodule formation in human bone marrow mesenchymal cells
  • Significantly decreases cell numbers in combination with sirolimus
Catalog No. 50-003-75942