Learn More
Medchemexpress LLC Purmorphamine (GMP) | 483367-10-8 | C31H32N6O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15108G100MG
Encompass_Preferred
Purmorphamine (GMP) is a version of Purmorphamine produced according to GMP guidelines. This GMP small molecule acts as an auxiliary reagent for cell therapy manufacturing and functions as a smoothened/Smo receptor agonist with an EC50 of 1 μM.
  • Functions as an auxiliary reagent for cell therapy manufacturing.
  • Acts as a smoothened/Smo receptor agonist with an EC50 of 1 μM.
  • Can generate embryoid bodies from human iPS cells and hESCs.
  • Converts hPSCs to NKX2-1 positive MGE cells.
  • Promotes ventralization of brain spheroids in differentiation process from hiPSCs.
  • Promotes hiPSC differentiation into functional neural tissue.
  • Induces hESCs differentiation into dopaminergic neurons.
Catalog No. 50-003-75864