Learn More
Selleck Chemical LLC Puromycin 2HCl
SDP
Supplier:  Selleck Chemical LLC S7417100MG
Encompass_Preferred
Puromycin 2HCl is an aminonucleoside antibiotic derived from Streptomyces alboniger It is an inhibitor of protein synthesis and has been used to study transcription regulatory mechanisms that control the sequential and coordinated expression of genes during cell differentiation
Catalog No. 50-000-03541