Learn More
Medchemexpress LLC Puromycin-bis(PEG2-amide)-Biotin | 2379889-82-2 | 98.8% | 1046.20 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1488655MG
Encompass_Preferred
Puromycin-bis(PEG2-amide)-Biotin is a binding molecule that targets ribosome-RNA complexes within single cells. It is specifically designed for use in single-cell translatome sequencing applications.
  • Binds to ribosome-RNA complexes within single cells.
  • Used for single-cell translatome sequencing.
  • Can penetrate single cells (mouse oocytes, zebrafish embryo cells, HeLa cells) at concentrations of 50-1000 μM for 30 seconds to 30 minutes.
Catalog No. 50-003-61639