Learn More
Medchemexpress LLC Puromycin-bis(PEG2-amide)-biotin | 2379889-82-2 | 99.9% | 1046.20 | C46H71N13O13S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14886510MG
Encompass_Preferred
Puromycin-bis(PEG2-amide)-Biotin is a biotin-conjugated puromycin derivative that binds the ribosome-RNA complex to label nascent peptides and facilitate affinity-based detection and purification of translating ribosomes. It is intended for use in translation profiling and sequencing workflows where capture or visualization of active translation is required.
  • Biotinylated puromycin analog for ribosome labeling.
  • Enables affinity capture of nascent peptides and ribosome-RNA complexes.
  • Suitable for single-cell translationome sequencing workflows.
  • High chemical purity (≈99.9%) and defined molecular weight.
  • Available in multiple package sizes, including 10 mg.
Catalog No. 50-002-85583