Learn More
TARGETMOL CHEMICALS INC Puromycin dihydrochloride 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T221950MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Puromycin dihydrochloride (CL13900 dihydrochloride) is a cinnamamide adenosine antibiotic and an inhibitor of protein synthesis. Puromycin dihydrochloride inhibits protein synthesis by binding to RNA and has antitumor and antitrypanosomal activity. Purity 99.86%
Catalog No. 50-287-9484