Learn More
Medchemexpress LLC HY-128972 25mg Medchemexpress, Purpurin 18 CAS:25465-77-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12897225MG
Encompass_Preferred
Medchemexpress, HY-128972 25mg Purpurin 18 CAS:25465-77-4 Purpurin 18, a derivative of chlorophyll, is an interesting dihydroporphyrin for generating photosensitizers. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1194