Learn More
Cayman Chemical Purvalnol B 5mg
SDP
Supplier:  Cayman Chemical 191155 MG
Encompass_Preferred
A CDK inhibitor that most potently inhibits Cdk2/cyclin A, Cdk2/cyclin E, Cdk5/p35, and Cdc2/cyclin B (IC50s = 6, 9, 6, and 6 nM, respectively); inhibits the growth of parasites, including Plasmodium, at micromolar doses
Catalog No. 50-298-1885