Learn More
Alkali Scientific Putrescine Dihydrochloride [1,4 Diaminobutane], 100 Grams
SDP
Supplier:  Alkali Scientific PUT001CP
Encompass_Preferred
Putrescine Dihydrochloride, also known as 1,4-Diaminobutane, is a key polyamine compound used in cell culture applications. It is essential for promoting cell growth and maintaining cellular viability in various culture systems. Putrescine is involved in the regulation of DNA synthesis, cell proliferation, and cellular differentiation. This reagent is used in laboratories to enhance cell culture conditions, particularly for the cultivation of mammalian cells. Available in a 100-gram quantity, Putrescine Dihydrochloride is utilized in studies of cell biology, cancer research, and tissue culture. It is an indispensable reagent for promoting optimal cell growth and ensuring stable experimental conditions.
Catalog No. 50-006-61188