Learn More
Leica Biosystems PV POLY-HRP M 110 DAB KIT
Supplier:  Leica Biosystems PV6113
Encompass
PowerVision Poly-HRP anti-Rabbit IHC Detection Systems are biotin-free, polymeric horseradish peroxidase (HRP) conjugated systems, for the manual detection of tissue-bound rabbit antibodies.
Catalog No. NC2581930