Learn More
Medchemexpress LLC PVA (Mw 145000, 99+% hydrolyzed, ~3300 polymerization) | 9002-89-5 | 99.1% | 145000 | 10 G
SDP
Supplier:  Medchemexpress LLC 10GHYY0850U4
Encompass_Preferred
PVA (Mw 145000, 99+% hydrolyzed, ~3300 polymerization) is a polyvinyl alcohol with a molecular weight of 145000 with hydrolysis properties. The hydrolysis degree refers to the conversion rate of the acetic acid group hydrolysis to the hydroxyl group in the original polyvinyl acetate. This polyvinyl alcohol is obtained from the polymerization of vinyl acetate to remove the acetic acid group by hydrolysis. Polyvinyl alcohol with different degrees of hydrolysis can be self-crosslinked to form frozen gels and can be used as biological excipients.
  • Molecular weight of 145000 (average)
  • Possesses hydrolysis properties
  • Obtained by polymerizing vinyl acetate
  • Can be self-crosslinked to form frozen gels
  • Used as biological excipients
  • Appears as a white to off-white solid
Catalog No. 50-003-64804