Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc PVA-SIL 5UM 120A 100X4.6MM
SDP

Supplier:  YMC America, Inc PV12S051046WT

Encompass_Preferred

Pva-Sil 5Um; 120A; 100 X 4.6mm

Catalog No. 50-900-05959


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.