Learn More
YMC America, Inc PVA-Sil grd 120A 5um 10x2.1mm
SDP
Supplier:  YMC America, Inc PV12S0501Q1GC
Encompass
YMC-Pack PVA-Sil is prepared from a 5 m 120 silica support which is bonded with a monomolecular polymer coating of vinyl alcohol The polymerized PVA completely covers both external and internal surfaces of the silica support protecting it against aggressive high pH buffers and solvents
Catalog No. 50-008-79533