Learn More
Perkin Elmer US LLC PVC Test Sample - Qty. 5
Supplier:  Perkin Elmer US LLC N5330340
Encompass_Preferred
The Measuring System requires the following calibration test samples to ensure optimal performance: PMMA, Steel, PVC, HDPE, Polycarb, PET Film, and G10 Epoxy. The test samples come standard with this DMA 8000 system, but they can also be ordered individually, as needed.
Catalog No. 50-211-1396