Learn More
Research Products International Corp PVDF MEMBRANE STRIPPING BUFFER 4X, 500 ML
SDP
Supplier:  Research Products International Corp P65000500.0
Encompass_Preferred
Classic formulation used for stripping primary and secondary antibodies from PVDF membranes. This method allows reprobing of the same membrane without having to run and blot multiple gels. Requires addition of B-Mercaptoethanol before using. Supplied in Molecular Grade Water.
Catalog No. 50-488-970