Learn More
Medchemexpress LLC PVR/CD155 Human Bi 20ug | 20ug
SDP
Supplier:  Medchemexpress LLC HYP7808420UG
Encompass_Preferred
The PVR/CD155 protein plays a key role in initiating natural killer (NK) cell effector functions by binding to CD96 and CD226 receptors These interactions form mature immune synapses that trigger adhesion lytic granule secretion interferon- (IFN- ) and cytotoxicity of activated NK cells PVR/CD155 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived PVR/CD155 protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-87577