Learn More
Graphic Controls LLC PW CIRCULAR CHARTS-100/PK,MIN5
Supplier:  Graphic Controls LLC 32021151
Encompass_Preferred
** Per Supplier requirements this item has a Minimum order quantity of 5 packs**PW-PARTLOW GC GC-7125532021151 - PW CIRCULAR CHARTS - 100 PER BXChart Size- 09.938"Time Rotation- 7DRanges- 0 - 50 Uniform along the Arc (0 TO 50 BY 5 )25 - 75 Uniform along the Arc (25 TO 75 BY 5 )Cross Reference:GC-71255To Fit Recorder Models:MRC 8000
Catalog No. NC2153315