Learn More
RS AMERICAS INC PWR SUPPLY ACDC 24VDC 5A 120W
Supplier:  RS AMERICAS INC 70936889
Encompass
Phoenix Contact 2904600 Power Supply, ACDC, 24VDC, 5A, 120W, DIN Rail Mount, QUINT POWER Series, GEN 4
Catalog No. NC3667686