Learn More
TARGETMOL CHEMICALS INC PX-12 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T228325MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PX-12 (PX12) (1-methylpropyl 2-imidazolyl disulfide) is a small-molecule inhibitor of Trx-1 (thioredoxin-1) stimulates apoptosis down-regulates HIF-1(alpha) and vascular endothelial growth factor (VEGF) and inhibits tumor growth in animal models. Since high levels of Trx-1 have been associated with colorectal gastric and lung cancers PX-12 is indicated as a potential cancer treatment in combination with chemotherapy for patients with advanced metastatic cancer. Initial trials correlated doses of Px-12 with increased patient survival. purity: 99%
Catalog No. 50-257-7178