Learn More
Medchemexpress LLC PXS-4681A | 1478364-87-2 | 99.4% | 296.75 | C10H14ClFN2O3S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11783350MG
Encompass_Preferred
A potent, selective, irreversible, and orally active semicarbazide-sensitive amine oxidase (SSAO/VAP-1) inhibitor intended for research use. Exhibits low-nanomolar potency across species and demonstrates anti-inflammatory effects in vivo. Supplied as a high-purity solid for biochemical and pharmacology studies.
  • Potent SSAO/VAP-1 inhibition with Ki ≈ 37 nM and IC50 values in the low nanomolar range.
  • High purity: 99.4%.
  • Molecular weight 296.75 g/mol and formula C10H14ClFN2O3S.
  • Irreversible mechanism of action and orally active in vivo models.
  • Suitable for biochemical assays and in vivo pharmacology; research use only.
  • Supplied as a 50 mg package.
Catalog No. 50-002-27256