Learn More
Medchemexpress LLC PXS-4787 hydrochloride | 2409964-40-3 | C10H13ClFNO2S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY151498A50MG
Encompass_Preferred
PXS-4787 hydrochloride is a specific and effective pan-LOX (lysyl oxidase) inhibitor that abolishes lysyl oxidase activity. It inhibits various LOX isoforms and reduces the deposition and crosslinking of collagen I secreted by human fibroblasts. This compound, intended for research use only, appears as a white to off-white solid with 99.71% purity and can be dissolved in DMSO at 100 mg/mL.
  • Specific and effective pan-LOX (lysyl oxidase) inhibitor
  • Abolishes lysyl oxidase activity
  • Inhibits various LOX isoforms (Bovine LOX, rh LOXL1, rh LOXL2, rh LOXL3, rh LOXL4)
  • Reduces deposition and crosslinking of collagen I
  • White to off-white solid with 99.71% purity
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-003-72316