Learn More
Selleck Chemical LLC PY-60 S9861-100MG
SDP
Supplier:  Selleck Chemical LLC S9861100MG
Encompass_Preferred
PY-60 is a small-molecule activator of the transcriptional coactivator Yes-associated protein (YAP) with EC50 of 1 6 M which acts by inhibiting a central scaffolding protein in the Hippo pathway Annexin A2 It promotes the regenerative repair of cutaneous wounds in pig and human models
Catalog No. 50-000-24734