Learn More
Medchemexpress LLC Py-maa-val-cit-pab-mmae | 2247398-68-9 | 98.5% | 1446.79 | C72H111N13O16S | 25 MG

Supplier: Medchemexpress LLC HY14887525MG
Py-MAA-Val-Cit-PAB-MMAE is a drug-linker conjugate used as a building block in antibody-drug conjugate (ADC) synthesis, combining a protease-cleavable linker with a cytotoxic payload for targeted oncology research. It is supplied as a white to off-white solid with supporting documentation for research use.
- High purity: 98.5%.
- Molecular weight 1446.79 and formula C72H111N13O16S.
- Solid, white to off-white appearance for easy handling.
- Documentation available: datasheet, certificate of analysis, and safety data sheet.
- Storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.