Learn More
Medchemexpress LLC Py-MAA-Val-Cit-PAB-MMAE | 2247398-68-9 | 98.5% | 1446.79 | C72H111N13O16S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1488755MG
Encompass_Preferred
Py-MAA-Val-Cit-PAB-MMAE is a drug-linker conjugate consisting of monomethyl auristatin E (MMAE) attached via a Py-MAA-Val-Cit-PAB cleavable peptide linker and intended for use in antibody-drug conjugate research and synthesis, including preparation of TRAILR2-targeting constructs.
  • Used as a building block for ADC synthesis.
  • Contains a Val-Cit-PAB cleavable peptide linker.
  • Includes monomethyl auristatin E payload for cytotoxic activity.
  • High purity suitable for preclinical research.
  • Available in small milligram quantities for synthesis workflows.
Catalog No. 50-002-03351