Learn More
ATCC Py8119; Breast Cancer; Mouse (Mus musculus)
Supplier:  ATCC CRL3278
Encompass
PY8119 is a mesenchymal-like cell line that was isolated in 2004 from the mammary gland of an adult female mouse with adenocarcinoma. This cell line was deposited by L Ellies.
Catalog No. 50-238-1891