Learn More
Medchemexpress LLC Pyclen dihydrochloride | 279.21 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY115853A
Encompass_Preferred
Pyclen dihydrochloride is a compound used for liver-specific MRI. Related compounds are synthesized as ligands of manganese (II) and have certain relaxivity and hepatocyte uptake ability. Pyclen dihydrochloride is a potential liver-specific contrast agent.
  • Used for liver-specific MRI.
  • Potential liver-specific contrast agent.
Catalog No. 50-004-37618