Learn More
Medchemexpress LLC Pyclen dihydrochloride | 78668-34-5 | 95.3% | 279.21 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY115853A100MG
Encompass_Preferred
Pyclen dihydrochloride is a compound utilized for liver-specific MRI. Related compounds have been synthesized as ligands of manganese (II) and possess certain relaxivity and hepatocyte uptake ability. It is considered a potential liver-specific contrast agent.
  • Used for liver-specific MRI
  • Related compounds are ligands of manganese (II)
  • Possesses certain relaxivity and hepatocyte uptake ability
  • Potential liver-specific contrast agent
Catalog No. 50-004-35721