Learn More
ABclonal Technology PYCR1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A27417100UL
Encompass_Preferred
This gene encodes an enzyme that catalyzes the NAD(P)H-dependent conversion of pyrroline-5-carboxylate to proline This enzyme may also play a physiologic role in the generation of NADP( ) in some cell types The protein forms a homopolymer and localizes to the mitochondrion Alternative splicing results in multiple transcript variants
Catalog No. 50-000-10000