Learn More
Abcam Pyocyanin, Catalase inhibitor, 5MG
Supplier:  Abcam AB1410995MG
Encompass_Preferred
MW 210.23 Da, Purity >98%. Catalase inhibitor. Aryl hydrocarbon receptor activator (Ki = 5.4 μM). Reduces gene expression of catalase. Promotes extracellular DNA release. Generates ROS. Induces apoptosis in neutrophils in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6903