Learn More
Apexbio Technology LLC Pyoluteorin 25683-07-2 5mg
SDP
Supplier:  Apexbio Technology LLC C40395
Encompass_Preferred
Pyoluteorin (CAS 25683-07-2) is a secondary metabolite exhibiting antibiotic and antifungal activities via inhibition of targeted pathogens including soilborne plant pathogens Pyoluteorin functions as a signaling molecule promoting its own biosynthesis and modulating secondary metabolite pathways in Pseudomonas fluorescens In vitro studies indicate pyoluteorin induces apoptosis in human triple-negative breast cancer MDA-MB-231 cells Current data do not report precise IC50 values for specific targets or models Based on its pharmacological profile pyoluteorin is applicable in research on microbial interactions control of plant pathogens investigation of bacterial communication and studies involving triple-negative breast cancer
Catalog No. 50-000-91538