Learn More
TARGETMOL CHEMICALS INC PYOLUTEORIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T217715MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pyoluteorin is an antibiotic that inhibits Oomycete fungi including the plant pathogen Pythium ultimum and suppresses plant diseases caused by this fungus. Pyoluteorin induces human triple-negative breast cancer MDA-MB-231 cells apoptosis in vitro. purity: 100%
Catalog No. 50-258-1006