Learn More
Selleck Chemical LLC PYR-41
SDP
Supplier:  Selleck Chemical LLC S712910MM/1ML
Encompass_Preferred
PYR-41 is the first cell-permeable inhibitor of ubiquitin-activating enzyme E1 with no activity at E2 PYR-41 induce apoptosis
Catalog No. 50-000-01315