Learn More
Medchemexpress LLC Pyr-Arg-Thr-Lys-Arg-AMC TFA | 1255501-99-5 | 99.5% | 941.95 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP4349A5MG
Encompass_Preferred
Pyr-Arg-Thr-Lys-Arg-AMC TFA is an AMC peptide. AMC is a decapeptide that proteases such as trypsin and thrombin specifically hydrolyze. This AMC peptide can be used to determine protease activity and the potency of enzyme inhibitors.
  • Can be used to determine the activity of proteases.
  • Can be used to determine the potency of enzyme inhibitors.
Catalog No. 50-003-97648