Learn More
Apexbio Technology LLC Pyrophosphatase Inorganic (E. coli) 50 units
SDP
Supplier:  Apexbio Technology LLC K108750
Encompass_Preferred
Pyrophosphatase Inorganic (E coli) purified from recombinant E coli strain carrying the E coli inorganic pyrophosphatase gene Inorganic pyrophosphatase (PPase) is a ubiquitous enzyme that catalyzes the hydrolysis of inorganic pyrophosphate into orthophosphate
Catalog No. 50-310-7413