Learn More
Apexbio Technology LLC [Pyr1]-Apelin-13 217082-60-5 1mg
SDP
Supplier:  Apexbio Technology LLC B70501
Encompass_Preferred
[Pyr1]-Apelin-13 (CAS 217082-60-5) is a potent and selective endogenous agonist of the Apelin receptor (APJ) By binding to APJ it activates downstream signaling pathways involved in the regulation of vascular tone and myocardial contractility In vitro studies demonstrate that [Pyr1]-Apelin-13 modulates biological processes pertinent to cardiovascular function This compound is widely utilized in research exploring the molecular mechanisms underlying cardiovascular diseases and is being investigated for its potential therapeutic relevance in this context
Catalog No. 50-000-87663