Learn More
Sigma Aldrich Fine Chemicals Biosciences (Pyr1)-Apelin-13 trifluoroacetate salt, 217082-60-5 (free base), MFCD01863147, 5 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SML20845MG
Encompass
Empirical Formula (Hill Notation): C69H108N22O16S · xC2HF3O2, Molecular Weight: 1533.80 (free base basis), ≥96% (HPLC), Synonym: 5-Oxo-L-prolyl-L-arginyl-L-prolyl-L-arginyl-L-leucyl-L-seryl-L-histidyl-L-lysylglycyl-L-prolyl-L-methionyl-L-prolyl-L-phenylalanine TFA, Pyroglutamated apelin-13, Pyroglutaminated apelin-13, Pyroglutamyl apelin-13, [Glp1]-Apelin-13, [Pyr-1]-apelin-13, [Pyr]-apelin-13, [pGlu1]-apelin-13, pERPRLSHKGPMPF TFA, pGlu-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe, TFA.
Catalog No. NC1609413