Learn More
Selleck Chemical LLC Pyr3 E4659-5MG
SDP
Supplier:  Selleck Chemical LLC E46595MG
Encompass_Preferred
Pyr3 is a selective and potent inhibitor of transient receptor potential canonical channel 3 (TRPC3) It inhibits TRPC3-mediated Ca2 influx in a dose-dependent manner with the IC50 value of 0 7 M It holds potential for treating TRPC3-related diseases like cardiac hypertrophy
Catalog No. 50-000-39437