Learn More
AdipoGen Pyraclonil
SDP
Supplier:  AdipoGen CDXP0179M010
Encompass_Preferred
Chemical. CAS 158353-15-2. Formula C15H15ClN6. MW 314.77. Synthetic. Nitrile herbicide used to control weeds in rice crops. Mode of action is through inhibition of protoporphyrinogen oxidase PPO.
Catalog No. 50-256-0209