Learn More
Medchemexpress LLC Pyraflufen-ethyl | 129630-19-9 | 98.0% | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY12692210MM/1ML
Encompass_Preferred
Pyraflufen-ethyl is a phenylpyrazole herbicide used for the control of annual and perennial broadleaf weeds.
  • Used for the control of annual and perennial broadleaf weeds
  • Functions as a phenylpyrazole herbicide
  • Acts as a metabolic enzyme/protease inhibitor
Catalog No. 50-003-42843