Learn More
AdipoGen Pyraflufen-ethyl
SDP
Supplier:  AdipoGen CDXP0169M010
Encompass_Preferred
Chemical. CAS 129630-19-9. Formula C15H13Cl2F3N2O4. MW 413.18. Synthetic. Phenylpyrazole herbicide derivative of pyraflufen. Used to control broad-leaved weeds and grasses in crops. Action of mode is through inhibition of protoporphyrinogen oxidase PPO.
Catalog No. 50-256-0212