Learn More
Medchemexpress LLC Pyrazine, 2,3,5,6-tetramethyl-, hydrochloride (1:x) | 76494-51-4 | 99.9% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN093510MG
Encompass_Preferred
Ligustrazine hydrochloride is the hydrochloride salt of tetramethylpyrazine provided as a high-purity research reagent for in vitro biological studies. It is reported to increase NOS and NO formation in cell models and is supplied for laboratory research use only (not for human consumption).
  • High reported purity suitable for research use.
  • Hydrochloride salt form increases water solubility compared with the free base.
  • Reported protective effects in cell-based assays, including increased NOS/NO activity.
  • Well-characterized chemical identifiers (CAS 76494-51-4; molecular weight ~172.65 g/mol).
  • Provided as an analytical/biological research chemical; follow safety data sheet guidance for handling.
Catalog No. 50-002-96093