Learn More
Medchemexpress LLC Pyrazine-2-amidoxime | 51285-05-3 | MFCD02258397 | 99.9% | 138.13 g/mol | C5H6N4O | 1 G

Supplier: Medchemexpress LLC HYW031471A1G
Pyrazine-2-amidoxime is a small-molecule synthetic intermediate and analogue of pyrazine-2-carboxamide with reported antimicrobial activity. It is supplied as a high-purity research chemical intended for use in synthetic and biological studies; follow supplier storage recommendations to maintain stability.
- High purity (99.9%)
- Available in multiple small pack sizes for research (250 mg-5 g)
- Suitable as a synthetic intermediate for medicinal chemistry
- Solid storage: 4°C, sealed, away from moisture
- In solvent: -80°C (6 months) or -20°C (1 month) when sealed
- Molecular formula C5H6N4O; molecular weight 138.13 g/mol
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.