Learn More
Medchemexpress LLC HY-122502 5mg Medchemexpress, Pyrazofurin CAS:30868-30-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1225025MG
Encompass_Preferred
Medchemexpress, HY-122502 5mg Pyrazofurin CAS:30868-30-5 Pyrazofurin, a pyrimidine nucleoside analogue with antineoplastic activity, inhibits cell proliferation and DNA synthesis in cells by inhibiting uridine 5'-phosphate (UMP) synthase. Pyrazofurin is an active, sensitive orotate-phosphoribosyltransferase inhibitor with IC50s between 0.06-0.37 µM in the three squamous cell carcinoma (SCC) cell lines Hep-2, HNSCC-14B and HNSCC-14C. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0988