Learn More
TARGETMOL CHEMICALS INC SP600125 200mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T3109200MG
Encompass_Preferred
SP600125 is a JNK1/2/3 inhibitor (IC50: 40/40/90 nM) and 10-fold higher selectivity than MKK4, 25-fold higher selectivity than MKK3, MKK6, PKCα, and PKB.
Catalog No. NC1624962